missing translation for 'onlineSavingsMsg'
Learn More

BAG3 Antibody (V65P1E8*D2) - BSA Free, Novus Biologicals™

Product Code. 18680517 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.03mL
0.10mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18680517 0.025 mL 0.03mL
18693068 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18680517 Supplier Novus Biologicals Supplier No. NBP2501120.025ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BAG3 Monoclonal antibody specifically detects BAG3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAG3
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone V65P1E8*D2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500
Formulation PBS
Gene Alias BAG family molecular chaperone regulator 3, BAG-3, BAG-family molecular chaperone regulator-3, Bcl-2-associated athanogene 3, BCL2-associated athanogene 3, BCL2-binding athanogene 3, Bcl-2-binding protein Bis, BIS, CAIR-1, Docking protein CAIR-1, MGC104307
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide MTDTPGNPAAP
Purification Method Protein A purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Endocrinology
Primary or Secondary Primary
Gene ID (Entrez) 9531
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.