missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAI1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-59011
This item is not returnable.
View return policy
Description
BAI1 Polyclonal antibody specifically detects BAI1 in Human, Bovine, Monkey samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| BAI1 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Bovine, Monkey | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.7 mg/mL | |
| ELISA 1:100 - 1:2000, Immunohistochemistry, Immunohistochemistry-Paraffin 10-15 μg/mL | |
| brain-specific angiogenesis inhibitor 1, FLJ41988, GDAIF | |
| Synthetic 18 amino acid peptide from N-terminus of human BAI1. | |
| 0.05 mg | |
| Angiogenesis, Apoptosis, Cancer, GPCR, Neuroscience | |
| 575 | |
| Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction