missing translation for 'onlineSavingsMsg'
Learn More

Baicalein, TRC

Product Code. 30134439
Change view
Click to view available options
Quantity:
250 mg
1 g
2.5 g
Unit Size:
1g
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134439 1 g 1g
30137206 2.5 g 2.5g
30010033 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134439 Supplier Toronto Research Chemicals Supplier No. TRCB1153001G
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 491-67-8
Molecular Formula C15 H10 O5
Molecular Weight (g/mol) 270.24
Synonym 4H-1-Benzopyran-4-one, 5,6,7-trihydroxy-2-phenyl-, Baicalein (6CI), Flavone, 5,6,7-trihydroxy- (7CI,8CI), 5,6,7-Trihydroxy-2-phenyl-4H-1-benzopyran-4-one, 5,6,7-Trihydroxyflavone, Baikalein, NSC 661431, Noroxylin
IUPAC Name 5,6,7-trihydroxy-2-phenylchromen-4-one
SMILES O=C1C=C(OC2=CC(O)=C(O)C(O)=C12)C=3C=CC=CC3

Specifications

Quantity 1 g
InChI Formula InChI=1S/C15H10O5/c16-9-6-11(8-4-2-1-3-5-8)20-12-7-10(17)14(18)15(19)13(9)12/h1-7,17-19H
Formula Weight 270.0528
Chemical Name or Material Baicalein
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.