Learn More
Avantor™ BAKERBOND Speedisk™ Extraction Disks
Product Code. 10406423 Shop All J.T.Baker Products
Change view
Click to view available options
Type:
Octadecyl (C18 XF) Extraction Disk
Octyl (C8) Extraction Disk
Unit Size:
Each
Pack of 20
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
10406423 Octadecyl (C18 XF) Extraction Disk Pack of 20
15568904 Octyl (C8) Extraction Disk Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10406423 Supplier Avantor™ Supplier No. 805606
  • ONLINE EXCLUSIVE PRICE
    577.00€
    508.18€ / Pack of 20
    Save 68.82€
    11% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket
Maximize capacity, adsorption and SPE speed. Avantor BAKERBOND Speedisk Extraction Disks are neither cartridge nor membrane that provides an excellent SPE tool for environmental samples and combines a thin bed of BAKERBOND sorbent with a special laminar configuration for testing drinking water, ground water, waste water, solid waste or process streams.
  • Pre-assembled with rigid support and baffle to maximize laminar flow, allow for filtration, resist clogging, and maximize capacity, adsorption, and speed
  • Require minimal glassware, set up time, and assembly
  • Minimizes impact of extractable compounds
  • Resists clogging and ensures high throughput rate
  • Reduce SPE run times
Specifications
For Use With (Application) Dirty Samples: EPA Methods 608,846, and Slightly Polar to Non-Polar Industrial Samples
For Use With (Equipment) Expanded Extraction Station (8095-06)
Sorbent C18 XF
Type Octadecyl (C18 XF) Extraction Disk
Quantity 20/Pk.
Diameter (Metric) Outer 50 mm