Learn More
Labconco Balance Vibration Isolator
Product Code. 15522495 Shop All Labconco Corporation Products
Change view
Click to view available options
Includes:
Marble slab, Four isolator pad, Protective top cover
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
15522495 Marble slab, Four isolator pad, Protective top cover Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15522495 Supplier Labconco Supplier No. 5234600P
  • ONLINE EXCLUSIVE PRICE
    1310.00€
    1111.65€ / Pack of 1
    Save 198.35€
    15% Off
Estimated Shipment: 24-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Isolator minimizes vibration to sensitive balances
  • Includes 13.4 W x 20.4 D x 2.7 in. thick marble slab, four isolator pads, and type 304 Stainless Steel protective top cover
  • Included with XPert Weigh Boxes
  • Applicable Products:
    • Precise Basic Glove Boxes
    • Precise Controlled Atmosphere Glove Boxes
    • Precise Hepa-filtered Glove Boxes
    • Protector Combination Glove Boxes
    • Protector Controlled Atmosphere Glove Boxes
    • Protector Filtered Glove Boxes
    • Protector Multi-hazard Glove Boxes (discontinued)
    • Xpert and Rxpert Filtered Balance Systems
    • Xpert Filtered Balance Stations
    • Xpert Weigh Boxes
Specifications
Product Type Vibration Isolator
Color White, Gray
Material Stainless Steel
Includes Marble slab, Four isolator pad, Protective top cover