missing translation for 'onlineSavingsMsg'
Learn More

Balsalazide disodium dihydrate, Thermo Scientific Chemicals

Product Code. 16564464 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16564464 5 g 5g
16554464 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Product Code. 16564464 Supplier Thermo Scientific Acros Supplier No. 465090050
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150399-21-6
Molecular Formula C17H17N3Na2O8
Molecular Weight (g/mol) 437.32
InChI Key HYSVUPBQPCRAJM-PIFVRXTCSA-L
IUPAC Name disodium (3Z)-3-(2-{4-[(2-carboxylatoethyl)carbamoyl]phenyl}hydrazin-1-ylidene)-6-oxocyclohexa-1,4-diene-1-carboxylate dihydrate
SMILES O.O.[Na+].[Na+].[O-]C(=O)CCNC(=O)C1=CC=C(N\N=C2\C=CC(=O)C(=C2)C([O-])=O)C=C1

Specifications

Quantity 5 g
Solubility Information Solubility aq. soln.: soluble. Other solubilities: sparingly soluble in methanol and ethanol, practically insoluble in organic solvents
Formula Weight 437.32
Chemical Name or Material Balsalazide disodium dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.