missing translation for 'onlineSavingsMsg'
Learn More

Balsalazide Disodium Dihydrate, TRC

Product Code. 30343143
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30343143 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30343143 Supplier Toronto Research Chemicals Supplier No. TRCB11629510MG
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Balsalazide Disodium Dihydrate
CAS 150399-21-6
Molecular Formula C17H13N3Na2O6.2(H2O)
Quantity 10 mg
Synonym 5-[(1E)-[4-[[(2-Carboxyethyl)amino]carbonyl]phenyl]azo]-2-hydroxyBenzoic acid, -, disodium salt, dihydrate (9CI), Benzoic acid, 5-[[4-[[(2-carboxyethyl)amino]carbonyl]phenyl]azo]-2-hydroxy-, disodium salt, dihydrate, (E)-, Balsalazide disodium dihydrate, Balsalazide disodium hydrate
InChI Formula InChI=1S/C17H15N3O6.2Na.2H2O/c21-14-6-5-12(9-13(14)17(25)26)20-19-11-3-1-10(2-4-11)16(24)18-8-7-15(22)23;;;;/h1-6,9,21H,7-8H2,(H,18,24)(H,22,23)(H,25,26);;;2*1H2/q;2*+1;;/p-2
SMILES O.O.[Na+].[Na+].Oc1ccc(cc1C(=O)[O-])N=Nc2ccc(cc2)C(=O)NCCC(=O)[O-]
IUPAC Name disodium;5-[(E)-[4-(2-carboxylatoethylcarbamoyl)phenyl]diazenyl]-2-hydroxybenzoate;dihydrate
Molecular Weight (g/mol) 437.3
Formula Weight 437.0811
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.