missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BANF1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-76751-0.025mg
This item is not returnable.
View return policy
Description
BANF1 Polyclonal specifically detects BANF1 in Human, Mouse, Rat, Hamster samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunofluorescence.
Specifications
| BANF1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| BAFMGC111161, barrier to autointegration factor 1, barrier-to-autointegration factor, BCRG1, BCRP1, Breakpoint cluster region protein 1, D14S1460 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 0.5-2 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 20 μg/mL | |
| AAH05942 | |
| BANF1 | |
| Antibody was raised against a 19 amino acid synthetic peptide from near the amino terminus of human BANF1. The immunogen is located within the first 50 amino acids of BANF1. Amino Acid Squence: TTSQKHRDFVAEPMGEKPV | |
| 0.025 mg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 8815 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction