missing translation for 'onlineSavingsMsg'
Learn More

BANP Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

168.00€

Specifications

Antigen BANP
Concentration 1 mg/mL
Dilution Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 10 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 10 μg/mL
Applications Western Blot
Classification Polyclonal
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18515053
View Documents
Novus Biologicals
NBP1-76295-0.025mg
0.025 mg
168.00€
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

BANP Polyclonal specifically detects BANP in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

BANP
Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 10 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 10 μg/mL
Polyclonal
Rabbit
Human
NP_001167014
54971
Antibody was raised against a 14 amino acid synthetic peptide near the center of human BANP. The immunogen is located within amino acids 300 - 350 of BANP. Amino Acid Squence: RTAWRRKQRGQSL
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
1 mg/mL
Western Blot
Unconjugated
RUO
PBS with 0.02% Sodium Azide
BEN domain containing 1, BEND1BEN domain-containing protein 1, BTG3 associated nuclear protein, Btg3-associated nuclear protein, DKFZp761H172, FLJ10177, FLJ20538, protein BANP, SMAR1Scaffold/matrix-associated region-1-binding protein, SMARBP1
BANP
IgG
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.