Learn More
Masterflex™ Barbed Elbow/tee/Y-connector Kits, Nylon
Organize the fittings at your workbench with these convenient fittings kits
Brand: Masterflex™ 31514-14
Additional Details : Weight : 0.05000kg
Description
- Designed for quick and easy assembly without tools.
- Combine these simple and economical fittings with flexible tubing.
- For use in low-pressure applications (generally less than 30psi).
Specifications
Barbed elbow/tee/Y-connector kits, Nylon, fittings kits (set of 1) | |
Barb Connection, Small | |
Nylon | |
Elbows: 1/16 in., 3/32 in., 1/8 in., 5/32 in., 3/16 in., 1/4 in., 5/16 in. Tees: 1/16 in., 3/32 in., 1/8 in., 5/32 in., 3/16 in., 1/4 in., 5/16 in. Y-connectors: 1/16 in., 3/16 in., 1/4 in., 3/8 in. |