Description
- These economical barbed fittings easily connect fluid paths without the use of tools.
- Just slip your flexible tubing or hose onto the barb to create a secure seal.
- Ideal for low-pressure applications.
- Chemically resistant.
- Available in a variety of sizes, materials, and configurations.
Specifications
Specifications
| Description | Barb x NPT(M) |
| Material | Polypropylene |
| Sterilization Method | Ethylene Oxide, Gamma |
| Product Type | Barbed NPT male pipe adapter elbows, Natural PP, 3/4 in. NPT x 3/4 in. |