missing translation for 'onlineSavingsMsg'
Learn More

Barium Acetate, 99.0 to 102.0%, ACS Reagent, Honeywell Fluka™

Product Code. p-7120206 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
Product Code. Quantity Packaging unitSize
15632630 100 g Poly Bottle 100g
15642630 500 g Poly Bottle 500g
2 options
This item is not returnable. View return policy

Product Code. 15632630

Brand: Honeywell-Fluka 243671100G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

ACS Reagent, 99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 543-80-6
Molecular Formula C4H6BaO4
Molecular Weight (g/mol) 255.42
MDL Number MFCD00012447
InChI Key ITHZDDVSAWDQPZ-UHFFFAOYSA-L
Synonym barium acetate, barium diacetate, acetic acid, barium salt, octan barnaty, barium di acetate, caswell no. 068a, octan barnaty czech, unii-fba31yj60r, ccris 7240, barium 2+ diacetate
PubChem CID 10980
IUPAC Name barium(2+);diacetate
SMILES [Ba++].CC([O-])=O.CC([O-])=O

Specifications

Color White
Quantity 100 g
Linear Formula (CH3COO)2Ba
UN Number NONH for all modes of transport
Beilstein 3693411
Formula Weight 255.42g/mol
Percent Purity 99.0 to 102.0%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Powder or crystals/granules
Density 2.46g/cm3
Chemical Name or Material Barium Acetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.