missing translation for 'onlineSavingsMsg'
Learn More

Barium Chloride Dihydrate, TRC

Product Code. 30344349
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30344349 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30344349 Supplier Toronto Research Chemicals Supplier No. TRCB1186805G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Barium Chloride Dihydrate
CAS 10326-27-9
Molecular Formula BaCl2H4O2
Quantity 5 g
Synonym Barium Chloride Dihydrate (8CI), Barium Chloride (BaCl2) Hydrate, Barium Dichloride Dihydrate
InChI Formula InChI=1S/Ba.2ClH.2H2O/h;2*1H;2*1H2/q+2;;;;/p-2
SMILES Cl[Ba]Cl
IUPAC Name dichlorobarium;dihydrate
Molecular Weight (g/mol) 244.26
Formula Weight 243.8641
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.