missing translation for 'onlineSavingsMsg'
Learn More

Barium Hydroxide, 8-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15557884 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15557884 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15557884 Supplier Avantor J.T.Baker Supplier No. 100601

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 12230-71-6
Physical Form Crystals
Quantity 500 g
Molecular Formula BaH18O10
MDL Number MFCD00149152
Synonym barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate
InChI Key ZUDYPQRUOYEARG-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++]
IUPAC Name barium(2+);dihydroxide;octahydrate
Molecular Weight (g/mol) 315.46
PubChem CID 17749109
Formula Weight 315.46
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 2.18kg
Chemical Name or Material Barium Hydroxide, 8-Hydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.