missing translation for 'onlineSavingsMsg'
Learn More

Barium Hydroxide Octahydrate, TRC

Product Code. 30333362
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30333362 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30333362 Supplier Toronto Research Chemicals Supplier No. TRCB1290005G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Barium Hydroxide Octahydrate
CAS 12230-71-6
Quantity 5 g
Molecular Formula BaH18O10
Synonym Barium Dihydroxide Octahydrate, Dihydroxybarium Octahydrate
InChI Formula InChI=1S/Ba.10H2O/h;10*1H2/q+2;;;;;;;;;;/p-2
SMILES O.O.O.O.O.O.O.O.O[Ba]O
IUPAC Name dihydroxybarium;octahydrate
Molecular Weight (g/mol) 315.46
Formula Weight 315.9952
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.