missing translation for 'onlineSavingsMsg'
Learn More

Barium iodide, ultra dry, 99.999% (metals basis), Thermo Scientific™

 

Brand:  Thermo Scientific Alfa Aesar 047118.14

Quantity 25g
Packaging Ampule

Product Code. 15482847

  • ONLINE EXCLUSIVE PRICE
    609.00€
    597.02€ / 25g
    Save 11.98€
    1% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Specifications

Specifications

13718-50-8
MFCD00003452
14,980
391.15
Odorless
Hygroscopic; Light sensitive
(metals basis)
25g
UN1564
Soluble in water,alcohol,and acetone.
99.999%
Ampule
5.15 g/mL
Barium iodide, Ultra dry
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.