Learn More
Euro Protection™ Barricade Tape
Product Code. 12048865
Change view
Click to view available options
Color:
Red, White
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
12048865 Red, White Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12048865 Supplier Euro Protection™ Supplier No. 70010
  • ONLINE EXCLUSIVE PRICE
    10.50€
    4.98€ / Pack of 1
    Save 5.52€
    52% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Dimensions (L x W) 100 m x 50 mm
Length (Metric) 100 m
Width (Metric) 50 mm
Color Red, White
Material HDPE
Type Barricade Tape