missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BarX1 Antibody (BARX1/2760), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08671AF350
This item is not returnable.
View return policy
Description
BarX1 Monoclonal antibody specifically detects BarX1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| BarX1 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| BarH-like homeobox 1, BARX homeobox 1, homeobox protein BarH-like 1 | |
| Recombinant full-length human BarX1 protein. (Uniprot: Q9HU1) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| BARX1/2760 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 56033 | |
| Store at 4°C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction