missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Basal Bodies of Cillia Antibody (LhS 28), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50114G
This item is not returnable.
View return policy
Description
Basal Bodies of Cillia Monoclonal antibody specifically detects Basal Bodies of Cillia in Human, Hamster samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| Basal Bodies of Cillia | |
| Monoclonal | |
| DyLight 488 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| LhS 28 | |
| 50 mM Sodium Borate | |
| BHK a21 Cytoskeletal preparation | |
| 0.1 mL | |
| Neuroscience | |
| Human, Hamster | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction