Learn More
BRAND™ Base for Plastic Bottles
Shop All Brand Products
Click to view available options
:
Diameter Outer: 94 - 96 mm
Specifications
Specifications
| Diameter (Metric) Outer | 96 mm |
| Capacity (Metric) | 1000 mL |
| For Use With (Equipment) | Automatic Burette Schilling, Plastic Bottles |
| Type | Base |