Learn More
Mecmesin™ Basic Force Gauge
Product Code. 11997979
Encompass
Change view
Click to view available options
Basic Force Gauge
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11997979 Basic Force Gauge
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
R\u00E9cupel-bijdrage uitgesloten
Product Code. 11997979 Supplier Mecmesin™ Supplier No. 853414
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Includes
Four nickel hydride batteries; Five mounting holes (57 mm pitch)
Designed for easy operation.
  • Provides tension and compression measurement
  • Calibration certificate traceable to national standards
  • Constructed in a rigid yet lightweight aluminum housing
  • Ergonomically attractive shape
  • Used as hand-held instrument
  • Allow testing under more controlled conditions
  • Accuracy: ±0.25%( full range)
Specifications
Product Type Gauge
Description Basic force gauge
Material Alumunum
For Use With (Application) Measure force