Learn More
Memmert™ Basic Series Water Baths
Product Code. 10738303 Shop All Memmert Products
Change view
Click to view available options
Capacity (Metric):
7 L
10 L
14 L
22 L
29 L
45 L
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10738303 7 L Each
10003832 45 L Each
10185702 22 L Each
10573693 14 L Each
10329342 10 L Each
10433302 29 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10738303 Supplier Memmert™ Supplier No. WNB7
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Corrosion-resistant stainless steel water bath. Memmert™ Basic Series Water Baths feature precise electronics and multiple temperature protection to guarantee the highest level of safety in the laboratory.
  • Microprocessor PID temperature controller
  • Digital display
  • Over-temperature protection
  • Integrated digital timer
Specifications
Controller Type Microprocessor PID
Capacity (Metric) 7 L
Certifications/Compliance CE, VDE, GS
Display Digital LED
For Use With (Application) For research laboratories, food and beverage, temperature control
Material Stainless Steel
Model WNB 7
Temperature (Metric) Maximum 95°C
Temperature Resolution ±0.1°C
Length (Metric) 468 mm
Length (Metric) Interior 240 mm
Width (Metric) 356 mm
Width (Metric) Interior 210 mm
Height (Metric) 238 mm
Height (Metric) Interior 140 mm
Depth (Metric) Interior 240 mm
Shipping Weight (Metric) 14 kg
Weight (Metric) 11 kg
Voltage 230 V
Wattage 1.2 kW
Frequency 50/60 Hz