Learn More
CleanSpace Technology™ Battery Charger
Product Code. 15970092 Shop All CleanSpace Technology Products
Change view
Click to view available options
Product Type:
Charger
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
15970092 Charger Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15970092 Supplier CleanSpace Technology™ Supplier No. PAF1101
  • ONLINE EXCLUSIVE PRICE
    97.98€
    76.00€ / Pack of 1
    Save 21.98€
    22% Off
 Restricted Item
Estimated Shipment: 21-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
The CleanSpace Battery Charger is specifically designed to re-charge the internal lithium polymer battery of the CleanSpace Power Units. The battery charger has two leads and interchangeable plugs for use in different countries.
  • Used with the revolutionary CleanSpace PAPR: light weight, no hoses or belts
  • Multinational interchangeable plugs
  • Dual leads for charging two PAPR power units simultaneously
Specifications
Product Type Charger
For Use With (Equipment) PAF-0034, PAF-0060, PAF-0070