missing translation for 'onlineSavingsMsg'
Learn More

Bazedoxifene N-Oxide, TRC

Product Code. 30006135
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006135 5 mg 5mg
30007942 10 mg 10mg
30009396 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006135 Supplier Toronto Research Chemicals Supplier No. TRCB1292805MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1174289-22-5
Molecular Formula C30 H34 N2 O4
Molecular Weight (g/mol) 486.6
Synonym 1H-Indol-5-ol, 1-[[4-[2-(hexahydro-1-oxido-1H-azepin-1-yl)ethoxy]phenyl]methyl]-2-(4-hydroxyphenyl)-3-methyl-, 1-[[4-[2-(Hexahydro-1-oxido-1H-azepin-1-yl)ethoxy]phenyl]methyl]-2-(4-hydroxyphenyl)-3-methyl-1H-indol-5-ol, Bazedoxifene N-Oxide
IUPAC Name 2-(4-hydroxyphenyl)-3-methyl-1-[[4-[2-(1-oxidoazepan-1-ium-1-yl)ethoxy]phenyl]methyl]indol-5-ol
SMILES Cc1c(c2ccc(O)cc2)n(Cc3ccc(OCC[N+]4([O-])CCCCCC4)cc3)c5ccc(O)cc15

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C30H34N2O4/c1-22-28-20-26(34)12-15-29(28)31(30(22)24-8-10-25(33)11-9-24)21-23-6-13-27(14-7-23)36-19-18-32(35)16-4-2-3-5-17-32/h6-15,20,33-34H,2-5,16-19,21H2,1H3
Formula Weight 486.2519
Chemical Name or Material Bazedoxifene N-Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.