missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCKDK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
415.00€ - 589.00€
Specifications
| Antigen | BCKDK |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18231980
|
Novus Biologicals
NBP2-54937 |
100 μL |
589.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18659606
|
Novus Biologicals
NBP2-54937-25ul |
25 μL |
415.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BCKDK Polyclonal specifically detects BCKDK in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| BCKDK | |
| Polyclonal | |
| Rabbit | |
| Protein Kinase | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 10295 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:TMESHLDTPYNVPDVVITIANNDVDLIIRISDRGGGIAHKDLDRVMDYHFTTAEASTQDPRISPLFGHLDMHSGAQSGPMHGFGFGLP | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| BCKDHKIN, branched chain alpha-ketoacid dehydrogenase kinase, branched chain ketoacid dehydrogenase kinase, EC 2.7.11, EC 2.7.11.4, mitochondrial | |
| BCKDK | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title