missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-6 Antibody (CL14611) [CoraFluor™ 1], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-26872CL1
This item is not returnable.
View return policy
Description
Bcl-6 Monoclonal antibody specifically detects Bcl-6 in Human samples. It is validated for Immunohistochemistry,Immunocytochemistry/ Immunofluorescence
Specifications
| Bcl-6 | |
| Monoclonal | |
| CoraFluor 1 | |
| B-cell CLL/lymphoma 6, B-cell lymphoma 6 protein transcript, BCL-5, BCL5B-cell lymphoma 5 protein, BCL-6, BCL6A, cys-his2 zinc finger transcription factor, LAZ3ZBTB27B-cell lymphoma 6 protein, lymphoma-associated zinc finger gene on chromosome 3, Protein LAZ-3, Zinc finger and BTB domain-containing protein 27, Zinc finger protein 51ZNF51, zinc finger transcription factor BCL6S | |
| This antibody was developed using a recombinant protein derived from P41182, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Tumor Suppressors | |
| 604 | |
| Store at 4⪚C in the dark. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| CL14611 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction