Learn More
Bcl rambo Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque:  Novus Biologicals NBP2-92603-0.1ml
 Afficher plus de versions de ce produit
Code produit. 18603992
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Voir les produits de remplacement »
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
Bcl rambo Polyclonal antibody specifically detects Bcl rambo in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Spécification
Spécification
Bcl rambo
Polyclonal
Western Blot 1:500 - 1:2000
Bcl2-L-13, BCL2-like 13 (apoptosis facilitator), bcl-2-like protein 13, Bcl-rambo, MIL1BCL-RAMBO, Protein Mil1
Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human BCL2L13 (NP_056182.2). MASSSTVPLGFHYETKYVVLSYLGLLSQEKLQEQHLSSPQGVQLDIASQSLDQEILLKVKTEIEEELKSLDKEISEAFTSTGFDRHTSPVFSPANPESSMEDCLAHLGEKVSQELKEPLHKALQMLLSQPVTYQAFRECTLETTVHASGWNKILVPLVLLRQMLLELTRRGQEPLSALLQFGVTYLEDYSAEYIIQQGGWGTVFSLESEEEEYPGITAEDSNDIYILPSDNSGQVSPPESPTVTTSWQSESLPVSLSASQSWHTESLPVSLGPESWQQIAMDPEEVKSLDSNGAGEKSEN
0.1 mL
Apoptosis
23786
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Western Blot
Unconjugated
PBS with 50% glycerol, pH7.3.