missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCL11A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11613-1-AP-20UL
This item is not returnable.
View return policy
Description
This is a rabbit polyclonal antibody raised against part chain of N-terminal BCL11A of human origin. There are three isoforms of BCL11A: isoform1 (773 aa), isoform2 (486 aa), and isoform3 (239 aa).
This gene encodes a C2H2 type zinc-finger protein by its similarity to the mouse Bcl11a/Evi9 protein. The corresponding mouse gene is a common site of retroviral integration in myeloid leukemia, and may function as a leukemia disease gene, in part, through its interaction with BCL6. During hematopoietic cell differentiation, this gene is down-regulated. It is possibly involved in lymphoma pathogenesis since translocations associated with B-cell maligcies also deregulates its expression. Multiple transcript variants encoding several different isoforms have been found for this gene.Specifications
| BCL11A | |
| Polyclonal | |
| Unconjugated | |
| BCL11A | |
| B cell CLL/lymphoma 11A, B cell lymphoma/leukemia 11A, BCL11A, COUP TF interacting protein 1, CTIP1, EVI 9, EVI9, HBFQTL5, KIAA1809, ZNF856 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14025, 305589, 53335 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9H165, Q9QYE3 | |
| BCL11A | |
| BCL11A Fusion Protein Ag2187 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction