missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCL7B Antibody (OTI2D9), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
256.00€ - 550.00€
Specifications
| Antigen | BCL7B |
|---|---|
| Clone | OTI2D9 |
| Dilution | Western Blot 1:500-2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30306655
|
Novus Biologicals
NBP3-44962-100ul |
100 μL |
550.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30306236
|
Novus Biologicals
NBP3-44962-30ul |
30 μL |
256.00€
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BCL7B Monoclonal antibody specifically detects BCL7B in Human,Mouse samples. It is validated for Western BlotSpecifications
| BCL7B | |
| Western Blot 1:500-2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| 9275 | |
| IgG1 | |
| >95% | |
| Antibody |
| OTI2D9 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| Allergen Hom s 3, B-cell CLL/lymphoma 7 protein family member B, B-cell CLL/lymphoma 7B | |
| Full length human recombiant protein of human BCL7B (NP_001698) produced in E.coli. | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title