Learn More
Berkshire International BCR™ Aluminium Flat Mop Handle
Product Code. 18899954 Shop All Berkshire Products
Change view
Click to view available options
Material:
Anodized Aluminum
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
18899954 Anodized Aluminum Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18899954 Supplier Berkshire International Supplier No. BCR.T2HANDLE.10
  • ONLINE EXCLUSIVE PRICE
    904.62€
    564.58€ / Pack of 10
    Save 340.04€
    37% Off
Estimated Shipment: 11-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Extremely manoeuvreable with a 360-degree swiveling head and telescopic handle extends from 95-180 cm.
  • Telescopic handle 95-180 cm
  • Anodized aluminum; lightweight and strong
  • Autoclavable (Recommended maximum of 121°C for 30 min)
  • Telescopic handle extends from 95-180 cm
Specifications
Product Type Cleanroom Mop Handle
Sterility Non-sterile
Autoclavable Autoclavable
Color-coded No
Material Anodized Aluminum