Learn More
BD BBL™ Cefinase™ β-Lactamase Detection Discs
Product Code. 11742974
Change view
Click to view available options
Quantity:
50 Discs
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11742974 50 Discs Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11742974 Supplier BD Supplier No. 231650
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Each disc is used to test a single bacterial strain for the presence of Β-lactamase
Rapid test used to isolate colonies of Neisseria gonorrhoeae, Staphylococcus sp., Haemophilus influenzae and anaerobic bacteria for the production of Β-lactamase:
  • Quarter inch disc impregnated with nitrocefin, a chromogenic cephalosporin
  • When a bacterium produces Β-lactamase in significant quantities, the yellow-colored disc turns red in the area where the isolate is smeared
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Nitrocefin
Content And Storage -20°C to +8°C
Packaging Type Disc
Quantity 50 Discs