missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Dehydrated Culture Media: Cary-Blair Transport Medium
Specifications
Specifications
| Certifications/Compliance | CCAM |
| Content And Storage | 2°C to 8°C |
| Form | Dehydrated |
| Media Type | Cary-Blair Transport Medium |
| Product Type | Cary-Blair Transport medium |
| Formulation | Sodium Thioglycollate 1.5g/L, Disodium Phosphate 1.1g/L, Sodium Chloride 5g/L, Agar 5g/L |
| For Use With (Application) | For collecting, transporting and preserving microbiological specimens, particularly Vibrio cholerae |
| Packaging Type | Bottle |
| Quantity | 500 g |
Safety and Handling
Product Identifier
- Cary and Blair Transport Medium
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- MIXTURE LIST-Contain: Sodium mercaptoacetate
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?