missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Taxo™ Urea Discs
Description
- 0.25 in. yellowish to slightly orange paper discs
- Discs contain sufficient amount of Urea R Broth to yield positive reaction with organisms producing sufficient urease to hydrolyze substrate
Order Info
50 discs.
Specifications
Specifications
| Antimicrobial Agents | Urea R Broth |
| Content And Storage | -20°C to +8°C |
| For Use With (Application) | Rapid detection of urease production |
| Quantity | 50 Discs |
For in vitro Diagnostic Use.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?