missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array (CBA) Human I-TAC; Flex Set, Bead A7
Description
Includes
Human I-TAC, Standard; PE Detection Reagent; Capture Bead A7.
- Bead-based immunoassay
- Human reactivity determined by testing samples with BD CBA Human I-TAC (CXCL11) Flex Set
Specifications
Specifications
| Antigen | I-TAC |
| Cross Reactivity | Human |
| Includes | Human I-TAC, standard, PE detection reagent, capture bead A7 |
| Quantity | 100 Tests |
| Content And Storage | 4°C |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human I-TAC flex set (bead A7) |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?