missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array (CBA) Mouse IL-13; Flex Set, Bead B8

Product Code. 15889318
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15889318 100 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15889318 Supplier BD Supplier No. 558349
 Restricted Item
Estimated Shipment: 17-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

Mouse IL-13 Standard (two vials), Mouse IL-13 PE Detection Reagent (one vial), Mouse IL-13 Capture Bead B8 (one vial)

Bead-based immunoassay for measuring mouse interleukin-13 (IL-13) in serum and cell culture supernatant samples

  • Mouse reactivity was determined by testing samples with Flex Set
  • Theoretical limit of detection is 2.4pg/mL
  • Storage buffer: Aqueous buffered solution containing BSA/fetal bovine serum along with other reagents depending on kit components

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-13
Cross Reactivity Mouse
Includes Mouse IL-13 standard (2 vials), Mouse IL-13 PE detection reagent (one vial), Mouse IL-13 capture bead B8 (one vial)
Quantity 100 Tests
Sample Type Serum, Plasma, Cell Culture Supernatant
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Product Type Mouse IL-13 flex set (bead B8)

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.