Learn More
BD Human Intracellular Cytokine Staining Starter Kit
Product Code. 15829698 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 Tests
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15829698 25 Tests 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15829698 Supplier BD Supplier No. 559302
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Includes
BD PharMingen Stain Buffer (FBS); Cytofix/Cytoperm Buffer; Perm/Wash Buffer; PE-conjugated anti-human or mouse IL-2; PE-conjugated anti-human or mouse IFN-g; PE-conjugated anti-human or mouse TNF-a; PE-conjugated isotype control cocktail; Purified blocking antibody cocktail; Leukocyte Activation Cocktail; MiCK-1 or HiCK-1 Cytokine Positive Control Cells; Staining Kit Manual
Serves as an introduction to the intracellular staining technique
  • Contains the essential reagents for the activation of a human peripheral blood cell population and their subsequent intracellular staining to detect IL-2, IFN-g-, and TNF-a-producing cells
TRUSTED_SUSTAINABILITY
Specifications
Detects IL-2, IFN-g- and TNF-a
Quantity 25 Tests
Content And Storage 4°C
Reactivity Human