Learn More
BD Microtainer™ MAP Microtube for Automated Process with K2EDTA and BD Microgard™ Closure
Product Code. 13224999
Change view
Click to view available options
Capacity (Metric):
250 to 500 μL
Additive:
1.0 mg K2 EDTA
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Additive Unit Size
13224999 250 to 500 μL 1.0 mg K2 EDTA Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13224999 Supplier BD Supplier No. 363706
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Designed for the collection, transport and automated processing of skin puncture samples from infants, children, geriatric, oncology and critical care patients
The BD Microtainer MAP tube is designed to improve turnaround time, safety and accuracy of capillary blood collection and testing in patients with limited blood volume such as infants, children, oncology patients and the elderly.
  • Streamlines operations by allowing for automated processing of your micro-collection hematology samples
  • Enhances patient safety by accommodating full-size patient identification labels at the time of collection
  • Improves workflow with one-piece design, no parts to assemble, no contaminated parts to discard
  • Latex-free, disposable and nonsterile
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Product Type MAP MicroTube for Automated Process
Additive 1.0 mg K2 EDTA
Sterility Non-sterile
Capacity (Metric) 250 to 500 μL
Closure Included Yes
Closure Color Lavender