Learn More
BD Mouse Hematopoietic Stem and Progenitor Cell Isolation Kit
Product Code. 15810249 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15810249 100 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15810249 Supplier BD Supplier No. 560492
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Includes
Monoclonal antibodies, isotype controls and other staining tools, flow cytometry compensation beads, protocols and analysis guidelines
For high purity isolation of hematopoietic progenitor and stem cells (HPCs and HSCs) from bone marrow
To perform multicolor flow cytometric cell sorting of hematopoietic progenitor cells (HPCs) and hematopoietic stem cells (HSCs) from mouse bone marrow samples
Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Formulation CD34 FITC, c-Kit PE, Sca-1 PE-Cy™7 and Lineage Cocktail APC, isotype controls, 7-AAD, BD™ CompBead compensation particles
For Use With (Application) Flow cytometry
Includes Monoclonal Antibodies, isotype controls and other staining tools, flow cytometry compensation beads, protocols and analysis guidelines
Quantity 100 Tests