missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beclin 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11306-1-AP-150UL
This item is not returnable.
View return policy
Description
Beclin 1 plays a central role in autophagy. It acts as a core subunit of the PI3K complex that mediates formation of phosphatidylinositol 3-phosphate. Different PI3K complex forms are believed to play a role in multiple membrane trafficking pathways - PI3KC3-C1 is involved in the initation of autophagosomes and PI3KC3-C2 is involved in the maturation of autophagosomes and endocytosis. Beclin 1 also plays a role in antiviral host defense.Specifications
| Beclin 1 | |
| Polyclonal | |
| Unconjugated | |
| BECN1 | |
| ATG6, Beclin 1, beclin 1, autophagy related, beclin1, BECN1, GT197, Protein GT197, VPS30 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114558, 56208, 8678 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O88597, Q14457, Q91XJ1 | |
| Becn1 | |
| Beclin 1 Fusion Protein Ag1843 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction