missing translation for 'onlineSavingsMsg'
Learn More

Beclomethasone, TRC

Product Code. 18092030
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18092030 25 mg 25mg
18021240 100 mg 100mg
17919959 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18092030 Supplier Toronto Research Chemicals Supplier No. TRCB13100025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4419-39-0
Molecular Formula C22 H29 Cl O5
Molecular Weight (g/mol) 408.92
Synonym Pregna-1,4-diene-3,20-dione, 9-chloro-11β,17,21-trihydroxy-16β-methyl- (7CI,8CI), (11β,16β)-9-Chloro-11,17,21-trihydroxy-16-methylpregna-1,4-diene-3,20-dione, 9-Chloro-11β,17,21-trihydroxy-16β-methyl-pregna-1,4-diene-3,20-dione, 9-Chloro-16β-methylprednisolone, 9α-Chloro-16β-methylprednisolone, Beclometasone, Beclomethasone, Becolvent
IUPAC Name (8S,9R,10S,11S,13S,14S,16S,17R)-9-chloro-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one
SMILES C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)CO

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C22H29ClO5/c1-12-8-16-15-5-4-13-9-14(25)6-7-19(13,2)21(15,23)17(26)10-20(16,3)22(12,28)18(27)11-24/h6-7,9,12,15-17,24,26,28H,4-5,8,10-11H2,1-3H3/t12-,15-,16-,17-,19-,20-,21-,22-/m0/s1
Formula Weight 408.1704
Percent Purity >95%
Chemical Name or Material Beclometasone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.