Learn More
Gilac™ Bedpan
Product Code. 17968601
Change view
Click to view available options
Volume (Metric):
2.5 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
17968601 2.5 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17968601 Supplier Gilac™ Supplier No. G101121
  • ONLINE EXCLUSIVE PRICE
    12.19€
    8.03€ / Pack of 1
    Save 4.16€
    34% Off
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This 2.5 L Bedpan is suitable for hospital use.
  • Suitable for hospital use.
  • Ergonomic and strong grip.
  • Outstanding durability even in the intensive use cycle.
  • Product is disinfected in a bedpan washer.
  • Researched product ergonomics ensure optimum cleaning.
  • Equipment at room temperature to avoid the cold sensation experienced with stainless steel equivalents.
Specifications
Product Type Bedpan
Color White
Material Polypropylene
Sterility Non-sterile
Volume (English) 0.7 gal.
Volume (Metric) 2.5 L