missing translation for 'onlineSavingsMsg'
Learn More

Beflubutamid D7 (benzyl D7), Dr. Ehrenstorfer

Product Code. 30314689
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30314689

Brand: Dr. Ehrenstorfer DREC10430100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Beflubutamid D7 (Benzyl D7)
Molecular Formula C18 D7 H10 F4 N O2
Quantity 10 mg
Synonym Beflubutamid D7 (Benzyl D7), 2-[4-Fluoro-3-(trifluoromethyl)phenoxy]-N-[(phenyl-d5)methyl-d2]butanamide, Beflubutamid-d7, (±)-Beflubutamid-d7 (benzylamine-d7), Benfluamid-d7, N-(Benzyl-d7)-2-(4-fluoro-3-(trifluoromethyl)phenoxy)butanamide
InChI Formula InChI=1S/C18H17F4NO2/c1-2-16(17(24)23-11-12-6-4-3-5-7-12)25-13-8-9-15(19)14(10-13)18(20,21)22/h3-10,16H,2,11H2,1H3,(H,23,24)/i3D,4D,5D,6D,7D,11D2
SMILES [2H]c1c([2H])c([2H])c(c([2H])c1[2H])C([2H])([2H])NC(=O)C(CC)Oc2ccc(F)c(c2)C(F)(F)F
IUPAC Name N-[dideuterio-(2,3,4,5,6-pentadeuteriophenyl)methyl]-2-[4-fluoro-3-(trifluoromethyl)phenoxy]butanamide
Molecular Weight (g/mol) 362.3698
Formula Weight 362.1635 g/mol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.