Learn More
behr Labor-Technik™ Graduated Cylinder
Product Code. 10361021
Change view
Click to view available options
Capacity (Metric):
500 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10361021 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10361021 Supplier behr Labor-Technik™ Supplier No. B00218164
Measuring cylinder 500 ml
Specifications
Material Glass
Capacity (Metric) 500 mL
Product Type Graduated Cylinder