Learn More
behr Labor-Technik™ behrotest™ Digestion Apparatus
Product Code. 10350011
Change view
Click to view available options
Capacity (Metric):
6 x 250 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10350011 6 x 250 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 10350011 Supplier behr Labor-Technik™ Supplier No. B00218105
  • ONLINE EXCLUSIVE PRICE
    9225.56€
    5905.64€ / Pack of 1
    Save 3319.92€
    35% Off
Estimated Shipment: 30-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
The digestion units of the model series behrotest™ InKjel are equipped with a high-performance infrared heater. Quality and position of the behr infrared heater guarantee the user identical heating phases and digestion temperatures at all sample positions.
  • Complete system with multi-level console, fume extraction unit, sample rack and glass digestion vessels
    • Rapid sample heating by means of direct heating via the infrared heater
      • Quartz infrared heaters reach a temperature of 830°C in 1 min
        • Various application possibilities: Rack for reaction vessels with 250 ml, 500 ml and 750 ml
          • With 500 ml and 750 ml vessels also suitable for nitrogen determination in water analysis
            • Easy conversion for various vessel sizes through the exchange of rack and fume extraction unit
              • Quick and problem-free digestion also for difficult samples: 25 freely configurable programs for block temperature and digestion time
                • behr one-button operation for extremely easy and quick programming
                  • Menu navigation in country language
Specifications
Product Type Programmable Infrared Digestion System
Description Programmable infrared digestion system for 6 glass vessels of 250 mL
Capacity (Metric) 6 x 250 mL
Depth (Metric) 0.8 m
Height (Metric) 0.64 m
No. of Places 6
Width (Metric) 0.55 m