Learn More
Bel-Art™ Protective Labeling Tape System: Tape Dispenser
Product Code. 10342671 Shop All SP Scienceware Products
Click to view available options
Color:
Silver
Unit Size:
Each
Product Code. 10342671
Brand: Bel-Art™ F134500000
Only null left
Add to basket
Add to basket
Utilizes a tape dispenser to fully cover and protect labels
  • Protective Labeling System provides labels that stand up to harsh laboratory conditions.
  • The clear tape protects labels from the freezer, autoclave, acids, alkalis and organic solvents.
  • Test results prove the tape remains securely affixed with no discoloration after being autoclaved for 40 minutes at 125°C (258°F), as well as after one week in liquid nitrogen at -196°C (-321°F).
  • All tape sizes are 1 mil (0.001 in.) thick with 1 mil (0.001 in.) of acrylic adhesive.
  • Durable tape dispenser is molded of strong ABS plastic with a steel serrated cutting blade; tape roll held in place by aluminum roller and spring-loaded cover
  • Optional epoxy-coated, weighted steel base reduces slippage on the benchtop when tape is dispensed; includes four nonslip rubber feet and screws
  • Clear, polypropylene tape has high resistance to chemicals and moisture; provides great versatility for heavy-duty lab use
  • White paper labels accept any kind of marker; perfect for use under the clear tape to label labware, bottles or samples
Specifications
Adhesive Type Non-adhesive
Color Silver
Display Consignment
Finish Gloss, Reflective
Format Roll
Material ABS
Printable Sides Top
Print Technology Ink Jet
Product Type Tape Dispenser
Sterility Non-sterile
For Use With (Application) Use to label containers, cabinets, drawers, books and lab equipment
Includes Tape dispenser, Steel cutting blade and Aluminum roller
Quantity 1 Ea.
Size 20.6 x 11.7 x 6 cm
Dimensions (LxW) 20.6 x 11.7cm
Length (English) 8.13 in.
Length (Metric) 20.65 cm
Width (English) 4.62 in.
Width (Metric) 11.7 cm
Height (English) 2.38 in.
Height (Metric) 6 cm