Learn More
Bel-Art™ Replacement Stopcocks
Description
Polypropylene body has Teflon™ PTFE plug, washer and bright red handle
- Three-way plug for vacuum draw, shut-off and vacuum release
- Taper fits on desiccator porte
Use on Desiccator Models: 08-594-15A, 08-594-16A, 08-594-15B, 08-594-16B, 08-594-15C, 08-594-16C, 01-817-50
Specifications
Specifications
| Type | 3-way Stopcock |
| Plug Bore | 0.24 in. x 0.38 in. O.D. |
| Material | Polypropylene |
| For Use With (Application) | Vacuum Desiccator Cabinets |
| For Use With (Equipment) | Scienceware™ desiccators, space saver and Techni-Dome™ 360 models |
| Includes | Washer, high visibility bright red handle with nut |
| Plug Material | PTFE |
| Outlet Port | Male Slip |
| Connectors | 3-way |
Teflon™ is a registered trademark of the Chemours Company FC LLC used under license by Bel-Arts Products, Inc.