Learn More
Bel-Art™ Secador™ Mini Desiccator Cabinet with Gas Ports
Product Code. 11872742
Change view
Click to view available options
Door Style:
Latched, swinging
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Door Style Unit Size
11872742 Latched, swinging Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11872742 Supplier Bel-Art™ Supplier No. F420751002
  • ONLINE EXCLUSIVE PRICE
    354.00€
    272.33€ / Pack of 1
    Save 81.67€
    23% Off
Estimated Shipment: 17-03-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The sturdy construction and compact size of this desiccator cabinet assures reliable storage for moisture sensitive products when benchtop space is at a premium.
  • Large side swinging door maximizes access to interior space.
  • Features patented seal design with tab latches and security loop for padlock or tamper-evident seals.
  • Opens right to left.
  • Top mounted gas ports with barbed fittings accept 0.25 in. I.D. tubing.
  • Easy-to-view dial hygrometer located in the door front.
  • Clear polystyrene for easy viewing of contents.
  • Gas ports can be closed to use desiccator with standard desiccant cartridges.
  • 10 D x 13.3 W x 14 in. H (25.4 x 33.8 x 35.6 cm).
  • Mini desiccator model is molded of polystyrene and does not block UV light.
  • For UV protection, select a Secador 1.0 model or larger.
Specifications
Color Clear
Cabinet Style Front-loading
Material Copolyester, Polystyrene
Product Type Desiccator
Door Style Latched, swinging
For Use With (Application) Assures reliable storage for moisture sensitive products when benchtop space is at a premium
Includes Fitting, stopcock (for 0.25 in. I.D. tubing) and instructions, top mounted two gas ports, locking loop with lock or tamper-evident seal, door-mounted dial hygrometer
Volume (English) 0.35 cu. ft.
Width (English) 13.3 in.
Width (English) Door 9.3 in.
Width (Metric) 33.8 cm
Width (Metric) Door 23.6 cm
Height (English) 8.5 in.
Height (English) Opening 5 in.
Height (Metric) 21.6 cm
Depth (English) 10 in.
Depth (Metric) 25.4 cm