Learn More
Bel-Art™ Sterileware™ Scoop An' Bag™ Sterile Sampler
Product Code. 11741566
Change view
Click to view available options
Material:
HIPS (FDA Grade)
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
11741566 HIPS (FDA Grade) Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11741566 Supplier Bel-Art™ Supplier No. H369100000
  • ONLINE EXCLUSIVE PRICE
    111.00€
    106.08€ / Pack of 50
    Save 4.92€
    4% Off
View all promotions
Estimated Shipment: 28-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Sample collection system includes a sterile 60 mL (2 oz.) sampling scoop and sterile 11.4 x 22.9 cm (4.5 x 9 in. ), 540 mL (18.3 oz.) volume polyethylene twirl-type bag with large label area to hold the sample.
  • One-piece scoop made from white, high-impact, FDA grade polystyrene plastic
  • Long handle keeps the hand away from the sample
  • Flat bottom allows scoop to remain upright for use as a weighing boat or to be placed on a flat surface
  • 50 individually sealed and sterilized wrapped sets per box ready for one time use and disposal sterile
  • Lot stamped for accurate lot tracing; shipped with a Certificate of Processing for Sterilization
Order Info
Sampling system comprised of a 2 oz. (60mL) white polystyrene scoop and a 9L x 4 0.50 in.W (228.6 x 114.3mm) polyethylene bag with a twirl-type closure packaged together.
Specifications
Certifications/Compliance FDA
Product Type Scoop Sampling System
For Use With (Application) Ideal for food, pharmaceutical and environmental sampling
Includes 60mL white polystyrene flat bottom one-piece sampling scoop with 12.74cm long handle and 7.8 L x 4.1 W x 2.9cm H bowl, 540mL 11.4 x 22.9cm polyethylene twirl-type bag with large label area to hold the sample
Material HIPS (FDA Grade)
Length (Metric) 20.5 cm
Sterility Sterile
Autoclavable Not Autoclavable
Shape Scoop