missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Belimumab Recombinant Monoclonal Antibody
GREENER_CHOICE

Recombinant Monoclonal Antibody

477.00€ - 1204.00€

Specifications

Antigen Belimumab
Concentration 1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30282839
GREENER_CHOICE
View Documents
Invitrogen™
MA558447
100 μg
477.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30284670
View Documents
Invitrogen™
MA558448
1 mg
1204.00€
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.

Belimumab is an intravenous immunosuppressant for the adjunctive treatment of systemic lupus erythematosus (SLE). More specifically, it is a fully human recombinant IgG1lambda monoclonal antibody produced from a recombinant NS0 cell line stably transfected with the belimumab heavy chain and light chain genes. It is the first biological treatment approved for the indication of SLE. Concomitant use with live or inactivated vaccines must be avoided. Belimumab was FDA approved on March 9, 2011. Belimumab consists of 2 heavy chains, and 2 light chains of the lambda subclass. Each heavy chain contains 452 amino acid residues and each light chain contains 214 amino acid residues. There are 3 post-translational modifications: a conserved N-linked glycosylation on the CH2 domain at Asn 303 of the heavy chain, the conversion of the N-terminal glutamine residue of the heavy chain into pyroglutamate, and loss of C-terminal lysine residue of the heavy chain.
Specifications

Specifications

Belimumab
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Lyophilized
IgG1
Human
hBlySmAb-1.1; LymphoStat-B
Antibody
1 mg/mL
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Unconjugated
Human
RUO
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Primary
Protein A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.