Learn More
Buerkle™ PVC Benchtop Draining Rack with Collecting Pan
Product Code. 11911611 Shop All Bürkle Products
Change view
Click to view available options
Positions:
31
49
73
Size:
400 x 400 mm
500 x 500 mm
600 x 600 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Positions Size unitSize
11911611 31 400 x 400 mm Each
11931611 49 500 x 500 mm Each
11921611 73 600 x 600 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11911611 Supplier Buerkle™ Supplier No. 96403040
Add to basket
Add to basket
Buerkle™ PVC Benchtop Draining Rack with Collecting Pan is easy and flexible to set down on a table or working surface.
  • Two large supports with wide bases ensure sufficient stability
  • The collecting pan can simply be removed for emptying
  • The positions of the stable rods can be changed at any time
  • Flexible, can be used on both sides
  • Draining rack PVC, rods PP
  • Incl. spanner for the rods
TRUSTED_SUSTAINABILITY
Specifications
Color Blue/White
Material PVC, Polypropylene
Positions 31
Size 400 x 400 mm
Width (English) 15.7 in.
Height (English) 15.74 in.
Includes Spanner, collecting pan, 5 x 60 mm rods, 26 x 100 mm rods, 2 x 150 mm rods
Product Type Benchtop Draining Rack
Height (Metric) 400 mm
Width (Metric) 400 mm